Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080735.1 | - | 4 | 791 | ||
BC014723.1 | - | 4 | 2156 | |||
BC082288.1 | - | 4 | 748 | 6490841 |