Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lifr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113386.1 | NP_001106857.1 | 3 | 536 | ||
NM_013584.2 | NP_038612.1 | 2 | 185 | |||
GenBank mRNA | AK155524.1 | - | 2 | 412 | ||
BC031929.1 | - | 2 | 178 | 4159053 | ||
BY716975.1 | - | 2 | 403 | |||
D17444.1 | - | 2 | 192 | |||
D26177.1 | - | 2 | 192 | |||
S73495.1 | - | 2 | 192 | |||
S73496.1 | - | 2 | 192 |