Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252563.1 | NP_001239492.1 | 4 | 857 | ||
NM_013592.4 | NP_038620.2 | 4 | 1122 | |||
GenBank mRNA | AJ006140.1 | - | 4 | 841 | ||
AK041073.1 | - | 4 | 830 | |||
AK160786.1 | - | 4 | 918 | |||
BC036558.1 | - | 4 | 857 | 1366191 |