Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322793.1 | NP_001309722.1 | 8, 9 | 1227 | ||
NM_001322794.1 | NP_001309723.1 | 8, 9 | 1110 | |||
NM_001322795.1 | NP_001309724.1 | 9, 10 | 1409 | |||
NM_001322796.1 | NP_001309725.1 | 8, 9 | 1391 | |||
NM_004969.3 | NP_004960.2 | 8, 9 | 1227 | |||
NR_136399.1 | 8, 9 | 1227 | ||||
XM_017016187.1 | XP_016871676.1 | 8, 9 | 1150 | |||
XM_017016188.1 | XP_016871677.1 | 8, 9 | 1744 | |||
XM_017016189.1 | XP_016871678.1 | 9, 10 | 1143 | |||
XM_017016190.1 | XP_016871679.1 | 8, 9 | 1270 | |||
GenBank mRNA | AK312810.1 | - | 8, 9 | 1152 | ||
BC096336.1 | - | 8, 9 | 1191 | 40008464 | ||
BC096337.1 | - | 8, 9 | 1191 | 40008465 | ||
BC096339.3 | - | 8, 9 | 1191 | 40008468 | ||
BX648462.1 | - | 8, 9 | 1392 | |||
M21188.1 | - | 8, 9 | 1208 |