Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004988.4 | NP_004979.3 | 3 | 1447 | ||
GenBank mRNA | BC017555.1 | - | 3 | 1414 | 3922845 |