Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pafah1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013625.4 | NP_038653.1 | 6 | 1080 | ||
GenBank mRNA | AK144908.1 | - | 6 | 1069 | ||
AK147207.1 | - | 6 | 1018 | |||
AK147229.1 | - | 6 | 1031 | |||
AK147237.1 | - | 6 | 1072 | |||
AK147309.1 | - | 6 | 1015 | |||
AK147507.1 | - | 6 | 1354 | |||
AK147515.1 | - | 6 | 1067 | |||
AK147533.1 | - | 6 | 1047 | |||
AK147793.1 | - | 6 | 742 | |||
AK164008.1 | - | 6 | 751 | |||
AK167448.1 | - | 6 | 1047 | |||
AK167824.1 | - | 6 | 1072 | |||
AK168929.1 | - | 6 | 720 | |||
AK168960.1 | - | 6 | 1035 | |||
AY189217.1 | - | 5 | 699 | |||
AY189218.1 | - | 5 | 699 | |||
BC014831.1 | - | 6 | 1019 | 4507474 | ||
BC026141.1 | - | 6 | 1044 | 4017963 | ||
BC046989.1 | - | 6 | 1052 | |||
BC099462.1 | - | 6 | 1018 | |||
L25108.1 | - | 6 | 980 | |||
L25109.1 | - | 2 | 176 | |||
U95116.1 | - | 6 | 1375 |