Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004990.3 | NP_004981.2 | 12, 13 | 1685 | ||
XM_006719398.3 | XP_006719461.1 | 11, 12 | 1520 | |||
GenBank mRNA | AK122956.1 | - | 13, 14 | 2233 | ||
AK222767.1 | - | 12, 13 | 1567 | |||
AK290857.1 | - | 12, 13 | 1556 | |||
AK293945.1 | - | 11, 12 | 1496 | |||
AK303349.1 | - | 8, 9 | 1189 | |||
BC002384.2 | - | 12, 13 | 1553 | 2961615 | ||
BC006328.2 | - | 12, 13 | 1553 | 4139330 | ||
BC011639.2 | - | 12, 13 | 1555 | 4120680 | ||
BC011849.2 | - | 12, 13 | 1553 | 4135154 | ||
BC015011.1 | - | 12, 13 | 1545 | |||
BT007338.1 | - | 12, 13 | 1547 | |||
D84224.1 | - | 12, 13 | 1570 | |||
X94754.1 | - | 12, 13 | 1554 | |||
Z49216.1 | - | 3, 4 | 281 |