Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244198.1 | NP_001231127.1 | 6 | 589 | ||
NM_001244200.1 | NP_001231129.1 | 7 | 801 | |||
NM_001244201.1 | NP_001231130.1 | 5 | 547 | |||
NM_001244202.1 | NP_001231131.1 | 5 | 1103 | |||
NM_013627.5 | NP_038655.1 | 6 | 1145 | |||
GenBank mRNA | AF443223.1 | - | 2 | 262 | ||
AJ292077.1 | - | 5 | 406 | |||
AK028059.1 | - | 5 | 548 | |||
AK032616.1 | - | 6 | 590 | |||
AK045805.1 | - | 5 | 1104 | |||
BC011272.1 | - | 6 | 833 | 4008490 | ||
BC036957.1 | - | 5 | 543 | 4504106 | ||
M77842.1 | - | 1 | 132 | |||
X63963.1 | - | 6 | 466 |