Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253877.1 | NP_001240806.1 | 11 | 1623 | ||
NM_001253878.1 | NP_001240807.1 | 11 | 1342 | |||
NM_001253879.1 | NP_001240808.1 | 10 | 1265 | |||
NM_001253880.1 | NP_001240809.1 | 8 | 980 | |||
NM_013827.3 | NP_038855.2 | 10 | 1261 | |||
GenBank mRNA | AB221622.1 | - | 7 | 668 | ||
AK030752.1 | - | 10 | 1211 | |||
AK031409.1 | - | 12 | 1330 | |||
AK032798.1 | - | 11 | 1584 | |||
AK032819.1 | - | 11 | 1585 | |||
AK137539.1 | - | 12 | 1570 | |||
AK139685.1 | - | 11 | 1343 | |||
BC024889.1 | - | 10 | 1233 | |||
BC029076.1 | - | 10 | 1172 | |||
BC050826.1 | - | 10 | 1215 | |||
BC057163.1 | - | 10 | 1058 | |||
BC071223.1 | - | 8 | 733 | |||
BC092237.1 | - | 10 | 1193 | 30637248 | ||
BC100340.1 | - | 8 | 911 | 30715034 | ||
S78454.1 | - | 7 | 665 |