Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142322.1 | NP_001135794.1 | 2 | 874 | ||
NM_001142323.1 | NP_001135795.1 | 2 | 874 | |||
NM_015742.2 | NP_056557.2 | 2 | 874 | |||
GenBank mRNA | AF143683.1 | - | 2 | 854 | ||
AK147347.1 | - | 1 | 64 | |||
AK148240.1 | - | 2 | 875 | |||
AK162587.1 | - | 2 | 975 | |||
BC138454.1 | - | 1 | 732 | 8861474 |