Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004998.3 | NP_004989.2 | 23 | 2870 | ||
GenBank mRNA | AB290177.1 | - | 23 | 2853 | ||
AK291688.1 | - | 23 | 2867 | |||
BC098392.1 | - | 23 | 2869 | 30527536 | ||
BQ054624.1 | - | 298 | ||||
U14391.1 | - | 23 | 2873 |