Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005001.4 | NP_004992.2 | 4 | 498 | ||
GenBank mRNA | AF050637.1 | - | 4 | 455 | ||
AF054178.1 | - | 4 | 469 | |||
BC003102.1 | - | 4 | 481 | 3537231 | ||
BC107892.1 | - | 4 | 469 | |||
CD172691.1 | - | 4 | 528 | |||
DA189271.1 | - | 4 | 476 |