Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017373.3 | NP_059069.1 | 2 | 598 | ||
GenBank mRNA | AK087541.1 | - | 2 | 599 | ||
AK145451.1 | - | 2 | 608 | |||
AY061760.1 | - | 1 | 270 | |||
BB872092.1 | - | 1 | 200 | |||
BC031377.1 | - | 2 | 442 | 5253195 | ||
BC100384.1 | - | 1 | 312 | 30479201 |