Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111324.2 | NP_001104794.1 | 4 | 883 | ||
NM_017464.5 | NP_059492.3 | 3 | 664 | |||
GenBank mRNA | AF009366.1 | - | 4 | 819 | ||
AK030985.1 | - | 4 | 817 | |||
AK033729.1 | - | 3 | 665 | |||
AK046357.1 | - | 3 | 627 | |||
AK054179.1 | - | 3 | 660 | |||
AK083374.1 | - | 4 | 811 | |||
BC053713.1 | - | 4 | 816 | 30077589 |