Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002011.3 | NP_001002011.2 | 8 | 1710 | ||
NM_001111102.2 | NP_001104572.1 | 8 | 1710 | |||
NM_019390.3 | NP_062263.1 | 8 | 1181 | |||
GenBank mRNA | AK003174.1 | - | 1 | 61 | ||
AK004619.2 | - | 8 | 1665 | |||
AK147150.1 | - | 8 | 1666 | |||
AK149998.1 | - | 8 | 1666 | |||
AK150391.1 | - | 8 | 1666 | |||
AK150501.1 | - | 8 | 1666 | |||
AK150624.1 | - | 8 | 1666 | |||
AK152539.1 | - | 8 | 1666 | |||
AK152646.1 | - | 8 | 1666 | |||
AK152846.1 | - | 8 | 1666 | |||
AK153295.1 | - | 8 | 1607 | |||
AK161221.1 | - | 8 | 1531 | |||
AK167858.1 | - | 8 | 1663 | |||
AK185150.1 | - | 2 | 104 | |||
AK208885.1 | - | 2 | 105 | |||
AK210233.1 | - | 2 | 105 | |||
AK216120.1 | - | 2 | 105 | |||
BC015302.1 | - | 8 | 1586 | 4240057 | ||
BC094020.1 | - | 8 | 1572 | 4187374 | ||
D13181.1 | - | 5 | 759 | |||
D14850.1 | - | 8 | 1181 | |||
DQ832702.1 | - | 8 | 1461 | |||
DQ832703.1 | - | 8 | 1461 | |||
X14170.1 | - | 8 | 1655 |