Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177369.1 | NP_001170840.1 | 15 | 1681 | ||
NM_001177370.1 | NP_001170841.1 | 14 | 1597 | |||
NM_019408.3 | NP_062281.1 | 14 | 1832 | |||
GenBank mRNA | AF155372.1 | - | 15 | 1759 | ||
AF155373.1 | - | 15 | 1796 | |||
AK134081.1 | - | 15 | 2100 | |||
AK137680.1 | - | 15 | 1671 | |||
AK148160.1 | - | 15 | 1669 | |||
AK150012.1 | - | 15 | 1660 | |||
AK153412.1 | - | 10 | 1241 | |||
BC027423.1 | - | 14 | 1598 | 3672913 | ||
DQ020182.1 | - | 14 | 1529 | |||
DQ020183.1 | - | 13 | 1650 | |||
DQ020184.1 | - | 12 | 1730 | |||
DQ020185.1 | - | 14 | 1529 |