Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019410.3 | NP_062283.1 | 3 | 910 | ||
GenBank mRNA | AJ272203.1 | - | 3 | 641 | ||
AK132651.1 | - | 3 | 718 | |||
AK164145.1 | - | 3 | 612 | |||
AK218126.1 | - | 1 | 270 | |||
BC024363.1 | - | 3 | 690 | 5059568 |