Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stmn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019641.4 | NP_062615.1 | 5 | 955 | ||
GenBank mRNA | AK002606.1 | - | 5 | 877 | ||
AK012231.1 | - | 5 | 877 | |||
AK013302.1 | - | 5 | 877 | |||
AK088251.1 | - | 5 | 877 | |||
AK088628.1 | - | 5 | 877 | |||
BC010581.1 | - | 5 | 864 | 3154091 | ||
BC031831.1 | - | 5 | 863 | 4500926 | ||
BC054396.1 | - | 4 | 856 | 6492045 |
Gene Symbol: Gm11223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030082.1 | - | 952 |