Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019724.3 | NP_062698.2 | 10 | 1757 | ||
GenBank mRNA | AB021228.1 | - | 10 | 1572 | ||
AF282844.1 | - | 10 | 1524 | |||
AK034828.1 | - | 3 | 180 | |||
AK048543.1 | - | 10 | 1637 | |||
AK077939.1 | - | 1930 | ||||
BC057926.1 | - | 10 | 1759 | 5292478 |