Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019724.3 | NP_062698.2 | 10 | 1937 | ||
GenBank mRNA | AB021228.1 | - | 10 | 1752 | ||
AF282844.1 | - | 10 | 1704 | |||
AK034828.1 | - | 3 | 360 | |||
AK048543.1 | - | 10 | 1818 | |||
AK077939.1 | - | 2110 | ||||
BC057926.1 | - | 10 | 1939 | 5292478 |