Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bloc1s6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019788.3 | NP_062762.1 | 2 | 215 | ||
GenBank mRNA | AK038381.1 | - | 2 | 193 | ||
AK077720.1 | - | 2 | 197 | |||
AK147116.1 | - | 2 | 218 | |||
AK160580.1 | - | 2 | 185 | |||
BY015259.1 | - | 2 | 216 |