Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ovol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019935.3 | NP_064319.1 | 4 | 1279 | ||
GenBank mRNA | AF134804.1 | - | 4 | 1295 | ||
AF134805.1 | - | 4 | 1261 | |||
AK003801.1 | - | 4 | 1314 | |||
AK028577.1 | - | 4 | 1280 | |||
AK132478.1 | - | 4 | 1280 | |||
AK132505.1 | - | 4 | 1318 | |||
AK132506.1 | - | 4 | 1314 | |||
BC021411.1 | - | 4 | 1359 | 5006712 |