Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020021.2 | NP_064405.2 | 1 | 900 | ||
GenBank mRNA | AK133089.1 | - | 1 | 900 | ||
BC137690.1 | - | 1 | 813 | 8860709 | ||
BC145745.1 | - | 1 | 813 | 40131048 |