Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039484.1 | NP_001034573.1 | 2 | 829 | ||
GenBank mRNA | AB039879.1 | - | 2 | 686 | ||
AF322631.1 | - | 2 | 638 | |||
AK044640.1 | - | 2 | 753 | |||
AK048864.1 | - | 2 | 832 | |||
AY374423.1 | - | 1 | 596 | |||
BC099932.1 | - | 2 | 772 | |||
BC141088.1 | - | 2 | 652 | 9055700 |