Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mvk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF137598.1 | - | 8 | 824 | ||
AK031084.1 | - | 8 | 1604 | |||
AK149630.1 | - | 9 | 921 | |||
AK177080.1 | - | 5 | 306 | |||
BC005606.1 | - | 9 | 877 | 3498140 |