Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025280.2 | NP_079556.1 | 2 | 207 | ||
GenBank mRNA | AK009429.1 | - | 2 | 239 | ||
AK132548.1 | - | 2 | 637 | |||
AK167740.1 | - | 2 | 211 | |||
BC028860.1 | - | 2 | 207 | 3673830 | ||
BC058169.1 | - | 2 | 236 | |||
X58472.1 | - | 2 | 212 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC099685.1 | - | 4497 | Chr. - - [-] on Build |