Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025281.3 | NP_079557.2 | 3 | 351 | ||
GenBank mRNA | AK006373.1 | - | 3 | 285 | ||
AK076727.1 | - | 3 | 356 | |||
AK211108.1 | - | 4 | 307 | |||
BC116923.1 | - | 2 | 207 | 40126242 | ||
BC116927.1 | - | 2 | 207 | 40126246 | ||
S60885.1 | - | 3 | 298 |