Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026810.2 | NP_081086.2 | 3 | 328 | ||
GenBank mRNA | AF250844.1 | - | 3 | 244 | ||
AK038265.1 | - | 3 | 321 | |||
AK145646.1 | - | 3 | 274 | |||
AK163861.1 | - | 3 | 262 | |||
AK168849.1 | - | 3 | 267 | |||
BC021815.1 | - | 3 | 282 | 4020489 |