Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252437.1 | NP_001239366.1 | 17 | 2766 | ||
NM_029657.4 | NP_083933.1 | 17 | 2763 | |||
GenBank mRNA | AK088533.1 | - | 17 | 2546 | ||
AK129161.1 | - | 13 | 2129 | |||
AK189949.1 | - | 1 | 105 | |||
AK194956.1 | - | 1 | 105 | |||
AK218167.1 | - | 1 | 105 | |||
BC046830.1 | - | 17 | 2612 | 6811606 |