Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080926.1 | NP_001074395.1 | 17 | 2456 | ||
GenBank mRNA | AJ312058.1 | - | 17 | 2302 | ||
AK030143.1 | - | 14 | 1966 | |||
AK038580.1 | - | 16 | 2493 | |||
BC127061.1 | - | 18 | 2209 | |||
D85463.1 | - | 18 | 2832 |