Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053214.2 | NP_444444.2 | 9, 10 | 1051 | ||
GenBank mRNA | AK158333.1 | - | 9, 10 | 966 | ||
BC046502.1 | - | 9, 10 | 1039 | 4167558 |