Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005027.3 | NP_005018.1 | 7 | 1415 | ||
NR_073517.1 | 7 | 1415 | ||||
GenBank mRNA | BC032647.1 | - | 6 | 1143 | 5518348 | |
BC070082.1 | - | 7 | 1387 | 30343107 | ||
BC090249.1 | - | 7 | 1347 | 6341766 | ||
X80907.1 | - | 6 | 1116 |