Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146583.1 | NP_666794.1 | 1 | 496 | ||
GenBank mRNA | AF102528.1 | - | 1 | 307 |