Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ei24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199494.1 | NP_001186423.1 | 3 | 341 | ||
NM_007915.5 | NP_031941.1 | 3 | 225 | |||
GenBank mRNA | AK145018.1 | - | 3 | 302 | ||
AK147776.1 | - | 3 | 254 | |||
AK159927.1 | - | 3 | 283 | |||
AK172233.1 | - | 3 | 305 | |||
BU148129.1 | - | 3 | 294 | |||
BY282890.1 | - | 3 | 342 | |||
U41751.2 | - | 3 | 229 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK139939.1 | - | 605 | Chr. - - [-] on Build |