Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ei24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199494.1 | NP_001186423.1 | 4 | 450 | ||
NM_007915.5 | NP_031941.1 | 4 | 334 | |||
GenBank mRNA | AK145018.1 | - | 4 | 411 | ||
AK147776.1 | - | 4 | 363 | |||
AK159927.1 | - | 4 | 392 | |||
AK172233.1 | - | 4 | 414 | |||
BU148129.1 | - | 4 | 403 | |||
U41751.2 | - | 4 | 338 |