Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx19a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007916.2 | NP_031942.2 | 12 | 1893 | ||
GenBank mRNA | AK003057.1 | - | 1 | 46 | ||
AK049467.1 | - | 12 | 1864 | |||
AK146548.1 | - | 12 | 1854 | |||
AK201912.1 | - | 1 | 220 | |||
BC011270.1 | - | 12 | 1865 | 3674608 | ||
L25125.1 | - | 12 | 1888 |