Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033578.2 | NP_001028750.1 | 6 | 941 | ||
NM_013257.4 | NP_037389.4 | 6 | 664 | |||
NM_170709.2 | NP_733827.2 | 6 | 664 | |||
GenBank mRNA | AF085233.2 | - | 5 | 455 | ||
AF169035.1 | - | 5 | 425 | |||
AK074666.1 | - | 6 | 618 | |||
AK223523.1 | - | 6 | 633 | |||
AK291428.1 | - | 6 | 941 | |||
BC015326.1 | - | 5 | 458 | 4391699 | ||
CR536581.1 | - | 3 | 203 | |||
CR933673.1 | - | 6 | 598 |
Gene Symbol: C8orf44-SGK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204173.1 | NP_001191102.1 | 8 | 1035 |