Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007924.2 | NP_031950.2 | 2 | 194 | ||
GenBank mRNA | AK159027.1 | - | 2 | 181 | ||
BC014816.1 | - | 2 | 164 | 4482339 | ||
BC024894.1 | - | 2 | 163 | 4159557 | ||
BY252808.1 | - | 2 | 178 | |||
BY272058.1 | - | 2 | 195 | |||
U80227.1 | - | 2 | 167 |