Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080388.2 | NP_001073857.1 | 6 | 1101 | ||
NM_001080389.2 | NP_001073858.1 | 6 | 1101 | |||
NM_001080390.2 | NP_001073859.1 | 6 | 818 | |||
NM_007928.3 | NP_031954.2 | 6 | 1101 | |||
GenBank mRNA | AK045329.1 | - | 6 | 675 | ||
AK154864.1 | - | 6 | 818 | |||
AK172444.1 | - | 6 | 1061 | |||
AK172664.1 | - | 6 | 744 | |||
AK215655.1 | - | 2 | 112 | |||
AK217185.1 | - | 1 | 61 | |||
AK220191.1 | - | 7 | 667 | |||
BC058556.1 | - | 6 | 1038 | 5289799 | ||
X70764.1 | - | 6 | 555 |