Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145961.1 | NP_001139433.1 | 2 | 311 | ||
NM_001145963.1 | NP_001139435.1 | 2 | 190 | |||
NM_001145964.1 | NP_001139436.1 | 2 | 181 | |||
NM_005072.4 | NP_005063.1 | 2 | 311 | |||
GenBank mRNA | AF047338.1 | - | 2 | 170 | ||
AF054505.1 | - | 2 | 170 | |||
AF054506.1 | - | 2 | 170 | |||
AK097808.1 | - | 2 | 177 | |||
AK293906.1 | - | 2 | 220 | |||
AK299042.1 | - | 2 | 97 | |||
AK302790.1 | - | 2 | 190 | |||
AK311073.1 | - | 2 | 183 | |||
AK316415.1 | - | 2 | 171 | |||
BC021193.2 | - | 2 | 239 | 3349710 | ||
BC035480.1 | - | 2 | 271 | |||
DC321966.1 | - | 2 | 312 | |||
DC333904.1 | - | 2 | 181 | |||
DC402650.1 | - | 2 | 190 | |||
U55054.1 | - | 2 | 225 |