Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evi5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007964.2 | NP_031990.2 | 3, 4 | 550 | ||
GenBank mRNA | AK134294.1 | - | 3, 4 | 751 | ||
AK165332.1 | - | 4, 5 | 543 | |||
AK165885.1 | - | 3, 4 | 503 | |||
AK192278.1 | - | 1, 2 | 40 | |||
AK216641.1 | - | 1, 2 | 226 | |||
BC030056.1 | - | 4, 5 | 557 | |||
BI737814.1 | - | 3, 4 | 474 | |||
U53586.1 | - | 3, 4 | 550 |