Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ezh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB004817.1 | - | 3, 4 | 240 | ||
AF483490.1 | - | 1, 2 | 129 | |||
AF483491.1 | - | 1, 2 | 129 | |||
AK045374.1 | - | 3, 4 | 228 | |||
AK129004.1 | - | 2, 3 | 201 | |||
AK138942.1 | - | 3, 4 | 223 | |||
AK140694.1 | - | 3, 4 | 247 | |||
AK154565.1 | - | 3, 4 | 220 | |||
AK164192.1 | - | 3, 4 | 239 | |||
BC007135.1 | - | 2, 3 | 200 | 3600884 | ||
U60453.1 | - | 3, 4 | 228 |