Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242368.1 | NP_001229297.1 | 5 | 752 | ||
NM_007972.4 | NP_031998.3 | 5 | 562 | |||
GenBank mRNA | AF087644.1 | - | 4 | 299 | ||
AJ222677.1 | - | 4 | 325 | |||
AK149449.1 | - | 4 | 346 | |||
AK154100.1 | - | 5 | 562 | |||
AK154572.1 | - | 5 | 679 | |||
AK170282.1 | - | 5 | 655 | |||
AK171096.1 | - | 5 | 562 | |||
BC003877.1 | - | 5 | 693 | 3495496 | ||
BC050219.1 | - | 5 | 738 | 30018889 | ||
CT010325.1 | - | 4 | 299 |