Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130409.1 | NP_001123881.1 | 4 | 578 | ||
NM_007982.2 | NP_032008.2 | 3 | 423 | |||
GenBank mRNA | AB011499.1 | - | 1 | 111 | ||
AB030035.1 | - | 1 | 111 | |||
AK005468.1 | - | 5 | 673 | |||
AK079821.1 | - | 4 | 675 | |||
AK220543.1 | - | 3 | 423 | |||
BC030180.1 | - | 4 | 508 | |||
CJ050166.1 | - | 4 | 536 | |||
JN971016.1 | - | 1 | 111 | |||
M95408.1 | - | 2 | 220 |