Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130409.1 | NP_001123881.1 | 12 | 1351 | ||
NM_007982.2 | NP_032008.2 | 11 | 1196 | |||
GenBank mRNA | AB011499.1 | - | 9 | 884 | ||
AB030035.1 | - | 9 | 884 | |||
AK079821.1 | - | 12 | 1448 | |||
AK220543.1 | - | 11 | 1196 | |||
BC030180.1 | - | 12 | 1281 | |||
JN971016.1 | - | 9 | 884 | |||
M95408.1 | - | 10 | 993 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK135507.1 | - | 1666 | Chr. - - [-] on Build |