Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007986.3 | NP_032012.1 | 4, 5 | 459 | ||
GenBank mRNA | AK051959.1 | - | 4, 5 | 362 | ||
AK136578.1 | - | 3, 4 | 449 | |||
BC019190.1 | - | 4, 5 | 363 | 5028579 | ||
Y10007.1 | - | 4, 5 | 459 |