Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007986.3 | NP_032012.1 | 25 | 2324 | ||
GenBank mRNA | BC019190.1 | - | 25 | 2228 | 5028579 | |
Y10007.1 | - | 25 | 2324 |