Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168357.1 | NP_001161829.1 | 3, 4 | 417 | ||
NM_005084.3 | NP_005075.3 | 3, 4 | 508 | |||
XM_005249408.4 | XP_005249465.1 | 3, 4 | 492 | |||
GenBank mRNA | AK290381.1 | - | 3, 4 | 467 | ||
AK294471.1 | - | 2, 3 | 290 | |||
BC038452.1 | - | 3, 4 | 417 | 5216829 | ||
BP326231.1 | - | 2, 3 | 369 | |||
U20157.1 | - | 3, 4 | 400 | |||
U24577.1 | - | 3, 4 | 455 |