Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081437.1 | NP_001074906.1 | 4 | 1558 | ||
NM_007992.2 | NP_032018.2 | 4 | 1558 | |||
GenBank mRNA | AK159361.1 | - | 4 | 1870 | ||
AK161522.1 | - | 4 | 1501 | |||
AK168688.1 | - | 4 | 1496 | |||
BC005443.1 | - | 4 | 1538 | 3490759 | ||
X75285.1 | - | 4 | 1458 |