Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008001.4 | NP_032027.2 | 2 | 837 | ||
GenBank mRNA | AK148151.1 | - | 2 | 838 | ||
BC011462.1 | - | 2 | 774 | 3674293 | ||
CN702035.1 | - | 2 | 327 | |||
U22325.1 | - | 2 | 436 |